missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ PolarSafe™ Cryogenic Storage Dewars with Racks

Cole-Parmer™ PolarSafe™ Cryogenic Storage Dewars with Racks safely store samples for up to 200 days without liquid nitrogen refill

Brand:  Cole-Parmer™ 4426057

 View more versions of this product

Product Code. 16639551

  • 1775.00€ / Each
Estimated Shipment: 08-08-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Liquid nitrogen dewars available in sizes from 2 L to 115 L
  • Ultra low evaporation loss enhances cryogen protection
  • Durable aluminum construction
  • Dual handles aid mobility
  • 5 year vacuum warranty
Specifications

Specifications

Static Holding Time: 76 Days
19.5 in.
4.875 in.
12.5 cm
Aluminum
35 L
49.53 cm
19.5 in.
49.53 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.