Learn More
Thermosafe™ PolarPack™ Refrigerant Gel Packs
Product Code. 12201372
Click to view available options
Weight (English):
8 oz./Pack
Unit Size:
Pack of 72
Product Code. 12201372
Brand: Thermosafe™ THV245020P
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Economical and practical refrigerant pack choice for shipping temperature-sensitive material such as food products, pharmaceuticals, and more. Thermosafe™ PolarPack™ Refrigerant Gel Packs are enclosed in durable polyethylene film.
  • Reliable non-mechanical refrigeration for the safe transport of food products, pharmaceuticals, and other medical products.
  • Multi-layered exterior minimizes surface condensation which protects product from moisture damage.
  • Outer film layer assures products arrive in top condition by absorbing condensation.
  • Phase change formulas ideal for products requiring frozen temperatures or extreme cold that may not tolerate dry ice or CO2.
  • Laminated, metalized film exterior provides radiant heat barrier, resists punctures.
  • Available in several sizes
  • Reusable
  • Non-toxic
TRUSTED_SUSTAINABILITY
Specifications
Quantity 72 Pk./Cs.
Weight (English) 8 oz./Pack
Dimensions (LxWxH) 6.5 x 3.5 x 0.75 in.