Learn More
Corning™ Pluronics Sacrificial Ink for Matribot™ Bioprinter
Product Code. 17760789 Shop All Corning Products
Click to view available options
:
Pluronics Sacrificial Ink; Quantity: 5 Syringes
Product Code. 17760789
Brand: Corning™ 6157
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Pluronics Sacrificial Ink is a ready-to-print sacrificial and support ink that exhibits excellent printability. Designed for use with Matribot™ Bioprinter.
  • Ready-to-print sacrificial and support ink that exhibits excellent printability
  • Easy to remove once printed by washing with cold PBS
  • Has been used for the fabrication of vascularized tissues, channels in microfluidic devices, and supporting scaffolds for complicated tissue constructs
  • Can also be diluted to 25% v/v with water to use as a learning tool to mimic dispensing of temperature-sensitive hydrogels
  • Designed for use with Corning™ Matribot™ Bioprinter
  • TRUSTED_SUSTAINABILITY
    Specifications
    Color Transparent
    For Use With (Equipment) Corning Matribot Bioprinter
    Product Type Sacrificial Ink
    Capacity (Metric) 2.7 mL/Syringe
    Description Sterile-A Quality
    Model Pluronics
    For Use With (Application) Fabrication of vascularized tissues, channels in microfluidic devices, and supporting scaffolds for complicated tissue constructs
    Quantity 5 Syringes
    Storage Requirements 4°C to 8°C