Learn More
asecos™ Plunger Can Sheet
Product Code. 17866536 Shop All asecos Products
Click to view available options
Capacity (Metric):
1 L
2 L
2.5 L
Material:
Stainless Steel
Steel
Unit Size:
Each
Product Code. 17866536
Brand: asecos™ 23135
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
The Plunger Can Sheet is ideal for safely moistening cleaning rags and sponges.
  • Incl. multilingual safety sticker for content labelling
  • Pumping unit with spring-mounted stainless steel dasher plate and integrated flame arrester
Specifications
Product Type Plunger Can
Capacity (English) 0.3 gal.
Capacity (Metric) 1 L
Color Gray
For Use With (Application) Safe moistening cleaning rags and sponges
Material Stainless Steel
Width (English) 5.20 in.
Width (Metric) 132.00 mm
Height (English) 10.16 in.
Height (Metric) 258.00 mm
Weight (English) 2.18 lb.
Weight (Metric) 0.99 kg