Learn More
Lauda™ Platinum Resistance Thermometers
Made from Platinum. Lauda™ Platinum Resistance Thermometers are for use with stainless steel version Pt100 to EN 60751, with Lemo connection.
Brand:  Lauda™ ETP 012
 View more versions of this product
Product Code. 10271602
  • 820.00€ / Each
Explore more special offers
Description
Description
  • Accuracy class A
  • Lemo connection
Specifications
Specifications
-200°C to +300°C
Platinum
150 mm