Learn More
Lauda™ Platinum Resistance Thermometers
Made from Platinum. Lauda™ Platinum Resistance Thermometers are for use with stainless steel version Pt100 to EN 60751, with Lemo connection.
Brand: Lauda™ ETP 012
This item is not returnable.
View return policy
Description
- Accuracy class A
- Lemo connection
Specifications
-200°C to +300°C | |
Platinum | |
150 mm |