Learn More
Platinum 13 wt% Rhodium wire, 0.203mm (0.008in) dia, ISA Type R Standard Grade Thermocouple, Thermo Scientific Chemicals
426.00€ - 1791.00€
Specifications
Chemical Name or Material | Platinum 13 wt% Rhodium wire |
---|---|
Name Note | 0.203mm (0.008 in.) dia., ISA Type R Standard Grade Thermocouple |
Avg. Mol. Wt. or Mol. Wt. Range | Pt:Rh; 87:13 wt% |
MDL Number | MFCD00198186 |
Odor | Odorless |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11380158
|
Thermo Scientific Alfa Aesar
042905.G1 |
1 m |
426.00€
1m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11390158
|
Thermo Scientific Alfa Aesar
042905.G5 |
5 m |
1791.00€
5m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum 13 wt% Rhodium wire | |
Pt:Rh; 87:13 wt% | |
Odorless | |
Wire |
0.203mm (0.008 in.) dia., ISA Type R Standard Grade Thermocouple | |
MFCD00198186 | |
≈0.64g/m |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only