Learn More
Kartell™ Plastilab™ Conical Stoppers
Click to view available options
Diameter (Metric) Bottom:
8.1 mm
10.4 mm
12.2 mm
16.4 mm
25.8 mm
Color:
Black
Blue
Brown
Green
Yellow
Standard Taper Joint:
NS 10/19
NS 12/21
NS 14/23
NS 19/26 Cone
NS 29/32
Unit Size:
Each
Pack of 10
Description
- Made from Polyethylene for tight and secure seal
- Raised embossed easy-to-read sizes
- Color-coded
Specifications
Specifications
| Certifications/Compliance | DIN 12242 |
| Description | Conical bottom |
| Standard Taper Joint | NS 29/32 |
| Color | Black |
| Diameter (Metric) Bottom | 25.8 mm |
| Material | Polyethylene |
| Product Type | Tapered Stopper |
| Shape | Pentagonal Grip |