Learn More
Microspec™ Plastic Inoculation Loops
Available as aseptic manufactured or ethylene oxide sterilized.
Brand:  Microspec™ C5
 View more versions of this product
Product Code. 16654822
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Includes 250 Plastic Inoculation Loops
  • Aseptic Manufactured or Ethylene Oxide Sterilized
  • 200 mm length
Specifications
Specifications
Single-use
Non-sterile
200 mm
Microseal Pouch
Not Autoclavable
1 μL
Blue
1 Ea.
Yes