Learn More
Sperian™ Honeywell™ Plastic RD40 Filters
Protects against a multitude of risks encountered in many environments. Sperian™ Honeywell™ Plastic RD40 Filters are positioned low to prevent obstruction in the field of vision.
17.00€ - 39.00€
Specifications
Cartridge Type | Filter |
---|---|
Connection Type | RD40 |
Product Type | Plastic Filters |
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
12653153
|
Sperian™
1788155 |
Gas vapor and particulates protection - A2B2E2K2 P3 large capacity for long duration work |
39.00€
Each |
In Stock
Log in to see stock. |
|||||
12613143
|
Sperian™
1786000 |
Dust and particulates protection |
17.00€
Each |
In Stock
Log in to see stock. |
|||||
12663153
|
Sperian™
1788161 |
Gas/vapor and particulates protection - AX large capacity for long duration work |
28.50€
Each |
Estimated Shipment: 24-07-2024
Log in to see stock. |
|||||
Description
- Body made of plastic provides optimum resistance
- Low breathing resistance
- Available in models to protect against airborne particulates, biohazards, chemicals, contamination and gas, vapors and smoke
Specifications
Filter | |
RD40 | |
Plastic Filters |