Learn More
Plas Labs™ Broome-Style Rodent Restrainers
Ideal for injections or blood sampling
Brand:  Plas Labs™ 553-BSRR
 View more versions of this product
Product Code. 13204979
  • 171.00€ / Each
Explore more special offers
Description
Description
  • Offer easy access to rodent
  • Prevent rodent from turning
  • Provide safety to the animal handler
  • Manufactured from clear materials to provide complete visibility of animal
Specifications
Specifications
Clear
Rodent Restrainer
Chemically sterilized or washed at 180°F
8 in.
2 in.
125 to 250 g
Injections or Blood Sampling
Noryl™ Plastic
Broome
20.32 cm
5.08 cm