Learn More
Terracycle™ Pipette Tip Boxes Zero Waste Box™
GREENER_CHOICE
Recycle Pipette Tip Boxes using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.
Brand:  Terracycle™ CS180-L-BE
 View more versions of this product
Product Code. 17845875
  • 266.38€ / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Description
Use this Zero Waste Box to recycle:
  • Empty pipette tip boxes
Do not send in:
  • Pipette tips, tubes and rigid lab plastics that are contaminated with materials considered hazardous or dangerous
  • Autoclaved material
  • Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people
  • Batteries, pressurized canisters, broken glass and medical waste
Important:
  • We do not collect any hazardous waste containing substances with toxic, reactive, corrosive, infectious, flammable, explosive properties or harmful to human health and the environment.
  • Please make sure that you do not mix hazardous waste with non-hazardous waste.
Specifications
Specifications
25 kg
Beige, Green, Red, White
Single-use
Large
Not Autoclavable
Recycling Box
55 lb.
38 x 38 x 97 cm
Yes
Non-sterile
Cardboard, Plastic