Learn More
Eppendorf™ Pipet Holder
Store up to six Eppendorf pipettes and/or liquid handling instruments in a comfortable and safe way. Eppendorf™ Pipet Holders are designed for use with the Eppendorf Research plus and Biomaster 4830.
Brand:  Eppendorf™ 3115600019
18.57 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.
Product Code. 10341104
  • 19.55€ / Each
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Description
  • Replacement pipet holder for pipette carousel
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Each