Learn More
Sartorius Pipet Charging Stand and Carousel
Shop All Sartorius Products
Click to view available options
Number of Channels:
1
4
Unit Size:
Each
Description
Includes
Universal AC-adaptor
For storing and charging Picus and eLINE mechanical and electronic pipets when not in use. Pipets should be stored in an upright position in order to avoid contamination from desks and benches.
Specifications
Specifications
For Use With | Pipette Tips |
Material | Polypropylene |
Number of Channels | 4 |
Type | Pipette Holder |
For Use With (Equipment) | Picus and eLINE Electronic Pipettes |
Includes | Universal AC adapter |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 7
Promotions
-
Multiple Manufacturers
Focus on Agri-Science Testing
Use promotion code 24136 to get amazing savings.Promo Code: 24136Expires: 2025-10-26
-
Multiple Manufacturers
Fisherbrand Sweet Deals on Top Products
Get up to 60% off select Fisherbrand productsPromo Code: 24168Expires: 2025-09-30
-
Multiple Manufacturers
New Offer Save Now
Product Content Correction
Product Title
Sartorius Pipet Charging Stand and Carousel >