Learn More
Brady™ Pipemarkers on a Card
European Standard Pipemarkers reinforce safety and aid maintenance.
Brand:  Brady™ N003288
 View more versions of this product
Product Code. 12529747
  • 11.85€ / Pack of 5
Estimated Shipment: 28-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols , if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, which saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
Specifications
Specifications
ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Permanent Rubber, Permanent, Pressure-sensitive
Card
Pipe Marker
Pipe and valve marking
French
5 Pk.
-40°C to +90°C
0.124 mm
150 mm
12 mm
Pipe Markers - Eau déionisée
White on Green
Polyester
Eau déionisée
5
5 Markers/Card
-40°F to +194°F
0.0049 in.
5.91 in.
0.47 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Special Offers
Special Offers