missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PIP4K2A, GST Tag Recombinant Protein
This PIP4K2A Recombinant Human Protein (full length) was expressed in insect cells.
Brand: Invitrogen™ A33576
This item is not returnable.
View return policy
Description
PIP4K2A lipid kinase or phosphatidylinositol-5-phosphate 4-kinase, type II, alpha, is a member of the phosphatidylinositol-5-phosphate 4-kinase family that catalyzes the phosphorylation of phosphatidylinositol-5-phosphate on the fourth hydroxyl of the myo-inositol ring to form phosphatidylinositol-4,5-bisphosphate. PIP4K2A plays an essential role in the phosphoinositide signal transduction cascades as the precursor to second messengers and is involved in the regulation of secretion, cell proliferation, differentiation, and motility. PIP4K2A may be one of the factors related to the regulation of the beta-globin gene expression and the different levels of Hb H in alpha-thalassemic patients.- Validated for use with the Adapta™ assay
- Available in multiple pack sizes for convenient use
Specifications
NP_005019.2 | |
72.5 kDa | |
PI3 Kinase | |
Human | |
GST Tag | |
Dry Ice | |
Full Length, Recombinant, Active |
Kinase Assay | |
100 μg | |
Full Length | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol | |
Insect | |
Liquid |