Learn More
OHAUS™ Pioneer™ Semi-Micro
Product Code. 15978606 Shop All Ohaus Products
Click to view available options
Model:
PX125D
PX125DM
PX225D
PX225DM
PX85
PX85M
Unit Size:
1 unit
Regulatory: This product is under eco-contribution tax.
Product Code. 15978606
Brand: OHAUS™ 30480169
Regulatory: This product is under eco-contribution tax.
OHAUS™ Pioneer™ Semi-Micro PX5 offers high accuracy and repeatability in laboratory, industrial and education applications. The PX5 is affordably priced, intuitively designed with a second line display for additional information, and USB and RS232 connectivity for easy communication.
  • The PX5 offers high accuracy and repeatability for essential weighing applications in laboratory, industrial and education settings at an economical price point.
  • Featuring a cast metal lower housing, sub-pan and stainless steel weighing pan, the PX5 is durably constructed for versatile, long-term use.
  • The PX5 features a second line display for additional information or guidance, a static removal bar for convenient grounding, and USB and RS232 connectivity for easy communication.
TRUSTED_SUSTAINABILITY
Specifications
Calibration InCal™ - Semi Automatic
Connection Type RS232 and USB device (included); GLP/GMP data output with real-time clock (except US version)
Capacity 82.00, 220.00 g
Display Type Backlit LCD, Second line display for additional information or guidance (dot matrix)
Readability 0.010000, 0.100000 mg
Languages English (US), French, German, Italian, Spanish (MX), Polish, Spanish
Linearity 0.000100 g
Model PX225DM
Repeatability 0.020000 mg, 0.100000 mg
Stabilization Time 10.000 sec.
Test Load 50 g, OIML E2 or ASTM CL 1
Weighing Modes Carat, Custom, Grain, Gram, Hong Kong Tael, Kilogram, Mesghal, Milligram, Momme, Newton, Ounce, Ounce Troy, Pennyweight, Pound, Singapore Tael, Taiwan Tael, Tical, Tola
Dimensions (L x W x H) 321 x 209 x 309 mm
Weight (Metric) 5.000 kg