missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sulfo-SBED Biotin Label Transfer Kit - Western Blot Application

Product Code. 10782345 Shop All Thermo Scientific Products
Click to view available options
Quantity:
8 Reactions
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 10782345

Brand: Thermo Scientific™ 33073

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Tag a purified protein to covalently crosslink and transfer biotinylation to its binding partner; then detect the results by Western blotting.

Thermo Scientific™ Pierce Sulfo-SBED Biotin Label Transfer WB Kit includes Sulfo-SBED Reagent to crosslink interacting proteins and transfer a biotin label from starting protein to target protein for detection by Western blotting.

Label transfer is a powerful in vitro method for protein interaction discovery. Sulfo-SBED Biotin Label Transfer Reagent can be used to identify previously unknown protein interaction binding partners and to more fully characterize the specific protein binding domains of other protein interactions. Sulfo-SBED is the abbreviation for Sulfo-N-hydroxysuccinimidyl-2-(6-[biotinamido]-2-(p-azido benzamido)-hexanoamido) ethyl-1,3'-dithioproprionate. It is a heterobifunctional chemical crosslinker capable of covalently attaching to primary amines at one end and to nearly any protein functional group at the other end. Unlike typical crosslinkers, Sulfo-SBED also includes a biotin group and a cleavable disulfide spacer arm. Together these features allow one to sequentially crosslink interacting proteins and transfer the biotin affinity tag from one protein (i.e., a purified “bait” protein) to another (possibly unknown “prey” protein).

Highlights:

  • Sulfo-NHS ester reacts with primary amines at pH 7-9
  • Aryl azide conjugates randomly when activated with UV light
  • Cleavable disulfide bond allows biotin label transfer
  • Discover protein interactions without radiolabeling

Includes:

Sulfo-SBED No-Weigh™ Format 8 x 1mg microtubes, BupH™ Phosphate Buffered Saline 1 pack, Label Transfer Buffer (20X) 200mL, Streptavidin-Horseradish Peroxidase Conjugate 0.1mg, Dithiothreitol (DTT) No-Weigh Format 8 x 7.7mg microtubes, Slide-A-Lyzer™ MINI Dialysis Units Plus Float 10kDa MWCO 10 units/pack

TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Quantity 8 Reactions
Product Type Crosslinker
Solubility Water
Content And Storage Sufficient For: 8 label transfer reactions for subsequent western blot analysis
• Sulfo-SBED, 8 x 1 mg
• PBS pack (makes 500 mL), 1 pack
• Label Transfer Buffer (20X), 200 mL
• Streptavidin-HRP Conjugate, 0.1 mg
• Dithiothreitol (DTT), 8 x 7.7 mg
• Slide-A-Lyzer MINI Dialysis Devices Plus Float, 10K MWCO, 10-device kit

Store at 4°C
Labeling Method Chemical Labeling, Label Transfer
Reactive Moiety Sulfo-NHS Ester, Aryl Azide
Product Line Pierce

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.