missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Premium Grade DSP (dithiobis(succinimidyl propionate))

Product Code. p-7000084 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 15376271

Brand: Thermo Scientific™ PG82081

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Optimize applications where product integrity and risk minimization are paramount with our highest quality formulation of this popular amine-reactive, cleavable crosslinker.

Optimize applications where product integrity and risk minimization are paramount with the specially characterized Thermo Scientific™ Pierce™ Premium Grade DSP, our highest quality formulation of this popular amine-reactive, cleavable crosslinker.

Thermo Scientific Pierce Premium Grade DSP is our highest quality formulation of this popular amine-reactive, cleavable crosslinker, specially characterized for applications where product integrity and risk minimization are paramount.

Compared to the standard grade product, Premium Grade DSP provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention and (d) change control notification. DSP is dithiobis(succinimidyl propionate), an amine-reactive crosslinker having NHS-ester reactive ends and a cleavable disulfide bond in its spacer arm. DSP contains an amine-reactive N-hydroxysuccinimide (NHS) ester at each end of an 8-carbon spacer arm. NHS esters react with primary amines at pH 7-9 to form stable amide bonds, along with release of the N-hydroxy-succinimide leaving group. Proteins, including antibodies, generally have several primary amines in the side chain of lysine (K) residues and the N-terminus of each polypeptide that are available as targets for NHS-ester crosslinking reagents.

Highlights:
  • DSP – a thiol-cleavable, homobifunctional and amine-reactive crosslinking agent
  • High quality – identity and purity confirmed by several tests, including quantitative NMR
  • Product integrity – enhanced level of testing and characterization compared to standard grade
  • Lot retention – ample supply of past lots retained to ensure future process testing
  • Change management – Change Control Notification (CCN) service
  • Consistent manufacture – batch-specific manufacturing documentation review
Applications:
  • Chemical crosslinking of intracellular proteins prior to cell lysis and immunoprecipitation
  • “Fix” protein interactions to allow identification of weak or transient protein interactions
  • Fix tissues for immunostaining without damaging RNA
  • Protein crosslinking to create bioconjugates via single-step reactions
  • Immobilize proteins onto amine-coated surfaces
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Amine
Cleavable By Thiols
Formulation Dithiobis(succinimidylpropionate); powder solid
Molecular Weight (g/mol) 404.42
PEGylated No
Sufficient For 2.47L of typical 1X (1mM) reaction solution
Spacer Arm Length 12.0 Å
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety NHS Ester
Spacer Medium (10 to 30 Å)
Form Powder
Quantity 1 g
Solubility DMF, DMSO
Format Standard, Single-use, Premium-grade
Water Soluble No
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.