missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NHS-PEG4-Azide

Product Code. 11894534 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 11894534

Brand: Thermo Scientific™ 26130

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Activated azido compounds to derivatize proteins or amine surfaces for selective conjugation, detection or purification by phosphine-modified probes.

Thermo Scientific™ Pierce NHS-Azide reagents are amine-reactive compounds that can be used to derivatize primary amines of proteins or amine-coated polymer surfaces for ligation to phosphine-modified molecules.

These NHS-ester compounds react to form covalent bonds with primary amines (e.g., side chain of lysine residues or aminosilane-coated surfaces). The azide (N3) group reacts with phosphine-labeled molecules by a mechanism known as Staudinger chemistry, enabling efficient and specific conjugation of derivatized molecules in biological samples. In combination with different phosphine-activated compounds, these NHS-Azide reagents facilitate a variety of labeling and crosslinking experimental strategies for investigating protein interactions and cellular pathways.

Highlights:

  • Soluble – reagents easily dissolve in water-miscible solvents for subsequent dilution in aqueous reaction mixtures with cell lysates and other biological samples

  • Compatible – reaction chemistry occurs effectively in simple buffer conditions; requires no accessory reagents such as copper or reducing agents

  • Specific – NHS esters are specific for covalent attachment to primary amines (side chain of lysine or N-terminus of polypeptides)

  •  

  • Chemoselective – azide and phosphine groups do not react or interfere with components of biological samples but conjugate to one another with high efficiency

  • Pegylated – choose the very short NHS-Azide or one of the two varieties whose spacer arms contain medium (4-unit) or long (12-unit) polyethylene glycol (PEG) groups for increased solubility and reach

TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Phosphine
Cleavable No
Description NHS-PEG4-Azide
Molecular Weight (g/mol) 388.37
PEGylated Yes
Spacer Arm Length 18.9 Å
Content And Storage Upon receipt store at -20°C protected from light and moisture.
Cell Permeability Not determined
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Amine, Azide, NHS Ester
Spacer Medium (10 to 30 Å)
Form Liquid
Quantity 100 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.