Learn More
Thermo Scientific™ Pierce™ LC-MS/MS System Suitability Standard (7 x 5 mixture)
Product Code. p-7228834 Shop All Thermo Scientific Products
Click to view available options
Concentration:
0.5 pmol/μL
5 pmol/μL
Unit Size:
100µL
25µL
Product Code. 16261851
Brand: Thermo Scientific™ A40010
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Pierce LC-MS/MS System Suitability Standard (7 x 5 mixture) contains seven HeavyPeptide AQUA Ultimate peptides provided at five dilutions distinguished by differential isotopic labeling to assess sensitivity and dynamic range of LC-MS/MS systems.
The Pierce LC-MS/MS System Suitability Standard (7 x 5 mixture) contains seven PRTC HeavyPeptide AQUA Ultimate peptides in five isotopologue versions for the generation of seven standard curves to assess sensitivity and dynamic range of LC-MS/MS systems.
The Pierce LC-MS/MS System Suitability Standard (7 x 5 mix) contains 7 of 15 peptides from the Pierce Peptide Retention Time Calibration Mixture (Cat. No. 88320). Each of the seven peptides has five versions (amino acids labeled with 0, 1, 2, 3, or 4 heavy isotopes) at five distinct concentrations. The 7 x 5 mix can be used with Skyline software to assess dynamic range, linearity, and lower limit of quantitation (LLOQ).
The Pierce LC-MS/MS System Suitability Standard (7 x 5 mixture) is:
High quality—contains five sets of seven HeavyPeptide AQUA Ultimate peptides, each distinguishable through unique isotopologue labeling
Optimized—enables scheduling of MS acquisition windows for improved quantification and increased multiplexing
Quantitative—assesses dynamic range of instrument and instrument sensitivity
Predictive—enables assessment of chromatography and MS instrument performance
Applications:
• Optimization of LC & MS parameters
• Standard for data-dependent acquisition (DDA)
• Standard for targeted MS (e.g., parallel-reaction monitoring (PRM)
• Identification of total peptide elution window
• Assessment dynamic range of nano or capillary flow LC-MS/MS systems
TRUSTED_SUSTAINABILITY
Specifications
Description Pierce™ LC-MS/MS System Suitability Standard (7 x 5 mixture) (0.5 pmol/μL)
Product Line Pierce
Shipping Condition Dry Ice
Concentration 0.5 pmol/μL
For Use With (Equipment) Mass Spectrometer
Type LC-MS/MS System Standard
Quantity 25 μL