missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Crossflow Filtration Cassettes, 3–100 kDa MWCO

Pierce Crossflow Cassettes are membrane-based devices designed for concentrating and diafiltration of sample solutions such as proteins, lipids, viruses, and cell culture supernatants with volumes from 0.50–3 L. Suited for manufacturing and pilot-scale tangential flow filtration processes.

Brand:  Thermo Scientific™ A57931

392.01 EUR valid until 2025-03-29
Use promo code "24111" to get your promotional price.


 View more versions of this product

Product Code. 17945086

  • 588.00€ / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Membrane Filter
Labscale TFF system
PES
2.5 nm
Certified Quality
Cassette
Polypropylene
10 kDa
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

  • Multiple Manufacturers
    Now extended - offer ends 28 March
    Promo Code: 24111

    Expires2025-03-28

    Days

    1

    Hours

    0

    Minutes

    30

    Seconds

    16
    New Offer
    Save Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Crossflow Filtration Cassettes, 3–100 kDa MWCO > 10 kDa MWCO, 1 cassette

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.