missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Phynomic C3 Cut Protection Gloves

Product Code. 17280381
Click to view available options
Size:
6
7
8
9
10
11
12
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Lightweight, sensitive gloves provide level 3/B cut resistance for mechanical activities. Aqua-polymer foam coating is breathable and enhances grip capabilities in dry or slightly damp applications.

  • Lightweight construction acts as a second-skin for tactile sensitivity
  • Level 3/B cut protection for mechanical tasks
  • Aqua-polymer foam coating is breathable and repels dampness
  • Gloves are made from materials that prevent excessive perspiration
  • Enhanced grip in dry, and slightly damp applications
  • 3D Ergo technology ensures a secure fit
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
  • Skin tolerance confirmed by the proDERM Institute for Applied Dermatological Research (11.0356-02, 11.0482-11)
TRUSTED_SUSTAINABILITY

Spezifikation

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material Aqua-Polymer Foam
Color Light Blue, Gray
Material Polyamide, Elastane, HPPE
Size 7
EN388 Standard 4X43B
For Use With (Application) Machine Operation, Ground Covering/Tilling
Gender Unisex
Series Phynomic C3
Yarn Gauge 15
Mehr anzeigen Weniger anzeigen
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'