missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHTPP, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective estrogen ERβ receptor antagonist that displays 36-fold selectivity over ERα. Exhibits full antagonism at ERβ in a cotransfection assay in human endometrial cancer cells (HEC-1), with minimal effects on ERα.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 423.31 |
Percent Purity | >99% |
Chemical Name or Material | PHTPP |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'