missing translation for 'onlineSavingsMsg'
Learn More

WTW™ pHotoFlex™ Portable Photometer with pH
GREENER_CHOICE

Product Code. 10435992
Click to view available options
Height (Metric):
117.3 mm
Unit Size:
Each
This item is not returnable. View return policy
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.

Product Code. 10435992

Brand: WTW™ 251100

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.

A unique multi-parameter function of photometry and electro-chemical pH measurement

With its unique multi-parameter function of photometry and electro-chemical pH measurement, pHotoFlex™ pH shows its strength with more complex tasks in environmental management, such as water framework guidelines, during process monitoring with changing sites and for fish hatcheries by determining NH3 and CO2: The energy-efficient LED optics with 6 wavelengths and a captive adapter for versatile applications with 16 and 28 cuvettes allows the measurement of the smallest concentrations with more than 180 programs for standard parameters and coloration. A list of favorites for the most commonly used tests, sample ID numbers and more than 3,000 measurements per battery set facilitate field measurements. The integrated pH function is a complete electro-chemical pH measurement of pH 0-16 with automatic buffer recognition (TEC/NIST) by using the comprehensive WTW selection of pH measuring chains. The temperature compensation automatically takes place in the admissible measuring ranges from -5 … 100 °C. The WTW MultiCal™ routine allows the automatic calibration with up to 3 calibration points. For field use, there is, for instance, the maintenance-free SenTix™ 41 is recommended, for precision measurements in the lab, the glass electrode SenTix™ 81 is frequently used. The optional PC software LSdata offers GLP-compliant data evaluation with data filter and allows the input of up to 100 user-defined programs, automatically calculate these as methods and save them to the pHotoFlex™. For the convenient use in the field, there is a case set available, including the pH probe SenTix™ 41 as a mobile lab with work surface and storage compartment as well as a lot of accessories, such as pH standards, tripod, pipettes and replacement batteries. For lab use, the photometer can be used in conjunction with a LabStation (optional) with site-supplied power and fitted with an external barcode reader.
TRUSTED_SUSTAINABILITY

Specifications

Display Type LED
For Use With (Application) Environmental Monitoring, Water Treatment, Beverage Industry, Wine Industry, Process Control, Multiparameter Applications for Photometry, pH and Turbidity
Height (English) 4.62 in.
Height (Metric) 117.3 mm
Length (English) 9.25 in.
Includes Approximately 200 programs and 100 user-defined programs, 1000 data sets and full electro-chemical pH functions
Length (Metric) 234.9 mm
Wavelength 436, 517, 557, 594, 610, 690nm
Width (Metric) 85.5 mm
Width (English) 3.37 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.