missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-BRD4 (Thr204) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA5117254
This item is not returnable.
View return policy
Description
Positive Samples: NIH/3T3 Immunogen sequence: ASTPP
BRD4, a ubiquitously expressed nuclear phosphoprotein, is a member of BET protein family. It is a human homolog of mouse MCAP and consists of 2 N-terminal bromodomains, a central ET-domain, and several kinase-like motifs in its N-terminus. It is expressed as 2 isoforms; the longer variant has a C-terminus region with a praline and glutamine-rich domains. BRD4 is a chromatin-binding protein whose expression is induced in response to growth stimuli. BRD4 acts at different stages of the cell cycle by interacting with proteins including RFC and SPA-1 and regulates cell growth. It also has a role in regulating chromosomal dynamics during mitosis. BRD4 functions as a cellular target for E2 protein of papillomavirus.Specifications
Phospho-BRD4 (Thr204) | |
Polyclonal | |
Unconjugated | |
BRD4 | |
Brd4; BRD4-NUT FUSION; Brd5; bromodomain containing 4; bromodomain-containing 4; bromodomain-containing 5; bromodomain-containing protein 4; bromodomain-containing protein 4; LOW QUALITY PROTEIN: bromodomain-containing protein 4; CAP; chromosome-associated protein; fsh-like protein; HUNK1; HUNKI; Mcap; mitotic chromosome-associated protein; Protein HUNK1; WI-11513 | |
Rabbit | |
Affinity chromatography | |
RUO | |
23476, 362844, 57261 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.97 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
O60885, Q9ESU6 | |
BRD4 | |
A synthetic phosphorylated peptide around T204 of human BRD4 (NP_4905971) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |