missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Phosphate Ortho/Total Cuvette Tests

Pre-dosed reagents for maximum safety. Easy to use and photometric evaluation, packaged in a fully equipped box. Officially approved for consent limits. Environmentally friendly due to less chemicals.

Brand:  Hach Lange™ LCK348


 View more versions of this product

Product Code. 10546015

  • 210.00€ / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Phosphate (Ortho/Total) cuvette test 0.05-1.5 mg/L PO4-P
  • For use with DR3900, DR6000, DR1900, DR2800, DR3800, DR5000 instruments
  • ISO 6878-1-1986, DIN 38405 D11-4 certifications
  • Shelf life of 23 months from production date
  • Storage conditions of 15 - 25 °C
  • Measuring ranges of 0.05 - 1.5 mg/L PO4-P and 0.15 - 4.5 mg/L PO4
  • Precise and reliable measurement values
  • Convenient and error-free dosing of the reagents
Specifications

Specifications

25
15°C to 25°C
Phosphate Ortho/Total Cuvette Test
0.5 to 5mg/L PO4-P
Phosphormolybdenum Blue
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Phosphat/Phosphate, DosiCapZip

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H302-Harmful if swallowed.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.