missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula

Used as a buffer system to maintain cell culture media in the physiological pH range. In combination with glucose, it provides the principal energy sources of cell metabolism. It is involved to irrigate, wash and dilute mammalian cells.

67.64€ - 123.62€

Specifications

Content And Storage Ambient temperatures
Packaging Type Plastic bottle
Form Liquid
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15440217
View Documents Promotion Details
Thermo Scientific Chemicals
J61917.AP
500 mL
67.64€
500mL
Please to purchase this item. Need a web account? Register with us today!
15450217
View Documents Promotion Details
Thermo Scientific Chemicals
J61917.K2
1 L
78.69€
1 L
Please to purchase this item. Need a web account? Register with us today!
15460217
View Documents
Thermo Scientific Chemicals
J61917.K3
2 L
123.62€
2 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula is used as a buffer system to maintain cell culture media in the physiological pH range. In combination with glucose, it provides the principal energy sources of cell metabolism. It is involved to irrigate, wash and dilute mammalian cells. It plays an important role in the transportation of tissue samples and as diluents in various reagent preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ambient temperatures
Plastic bottle
Liquid
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.