missing translation for 'onlineSavingsMsg'
Learn More

Phenyl trifluoroacetate, 98%, Thermo Scientific Chemicals

Product Code. 11419217
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500-73-2
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.121
MDL Number MFCD00039232
InChI Key DVCMYAIUSOSIQP-UHFFFAOYSA-N
Synonym phenyl trifluoroacetate, acetic acid, trifluoro-, phenyl ester, acetic acid, 2,2,2-trifluoro-, phenyl ester, trifluoroacetic acid phenyl ester, phenyltrifluoroacetate, acmc-209kjz, dvcmyaiusosiqp-uhfffaoysa
PubChem CID 68143
IUPAC Name phenyl 2,2,2-trifluoroacetate
SMILES C1=CC=C(C=C1)OC(=O)C(F)(F)F

Specifications

Density 1.276
Boiling Point 146°C to 147°C
Flash Point 52°C (127°F)
Refractive Index 1.423
Quantity 5 g
UN Number UN3272
Sensitivity Moisture sensitive
Formula Weight 190.12
Percent Purity 98%
Chemical Name or Material Phenyl trifluoroacetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'