Learn More
Phenanthrene-4,5-dicarboxylic acid, 97%, Thermo Scientific™
Product Code. 15471078
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 15471078
Brand: Thermo Scientific Alfa Aesar H54939.03
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5462-82-8
Molecular Formula C16H10O4
Molecular Weight (g/mol) 266.252
MDL Number MFCD11110581
InChI Key RAGXPRCIICFJIK-UHFFFAOYSA-N
Synonym 4,5-phenanthrenedicarboxylic acid, phenanthrene-4,5-dicarboxylate, 4,5-phenanthrenedicarboxylicacid, phenanthrene-4,5-dicarboxylic acid
PubChem CID 95075
ChEBI CHEBI:81603
IUPAC Name phenanthrene-4,5-dicarboxylic acid
SMILES C1=CC2=C(C(=C1)C(=O)O)C3=C(C=CC=C3C(=O)O)C=C2
Specifications
Melting Point 244°C to 246°C
Quantity 1 g
Formula Weight 266.25
Percent Purity 97%
Chemical Name or Material Phenanthrene-4,5-dicarboxylic acid