missing translation for 'onlineSavingsMsg'
Learn More

Christeyns™ Phago'Spray DASR 70

Phago'Spray DASR 70 is a hydroalcoholic disinfectant for the disinfection of hands, surfaces, equipment and furniture.

66.00€ - 66.85€

Specifications

Product Type Alcohol-based Surface Disinfectant
Color-coded No
Form Clear Liquid
pH Neutral
Sterility Non-sterile
View More Specs

Products 2
Product Code Brand Scent Size Price Quantity & Availability  
Product Code Brand Scent Size Price Quantity & Availability  
16475940
View Documents Promotion Details
Christeyns™
65049
Fresh Scent 750 mL
66.85€
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
16698472
View Documents Promotion Details
Christeyns™
C65050
Unscented 5 L This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
Description

Description

  • Can be used in application on a single-use cloth for the rapid disinfection of small furniture, trolleys, external surfaces of non-invasive medical devices (operating table, operating light, monitors, cables and connectors, stethoscopes, reflex, blood pressure monitors) and small surfaces close to the patient
  • Can be used, without rinsing, on surfaces that may be in direct contact with foodstuffs (table, worktop, trays, meal trolley, slicers, choppers, knives, etc.)
  • Broad antimicrobial spectrum from 30 sec.
  • Food contact, without rinsing
  • No streaks or residue on drying
  • Ecocertified
Specifications

Specifications

Alcohol-based Surface Disinfectant
Clear Liquid
Non-sterile
No
Neutral
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Product Certifications