Learn More
SI Analytics™ PH-METER Set HandyLab 680 Science
The HandyLab 680 pH meter set with pHT-combination electrode A162 IDS and Z389 for demanding applications allows you to measure any two parameters simultaneously; pH, ORP, conductivity or oxygen.
Brand:  SI Analytics™ 285204690
Product Code. 16379255
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Portable pH IDS meter
  • Two connectors for simultaneous measurement of pH, LF and DO
  • IDS technology
  • Auto-read function
  • CMC (Continuous measurement control)
  • QSC (Quality sensor control)
  • Traceability of the measuring values
  • User administration
  • Transmission of all data, in .csv format
  • Colour graphic display
  • Waterproof design IP67
TRUSTED_SUSTAINABILITY
Specifications
Specifications
USB, USB Stick
1-, 2-, 3-, 4-, 5-points
-5°C to +105°C
0.00 Ohm/cm to 100 Mohm/cm ±0.5% of mean value
0 to 1999 mg/L, 0 to 199.9 g/L ±0.5% of mean value
0 to 14
±1200.0, ±0.2
0.00 to 2000 mS/cm ±0.5% of mean value
0.0 to 70.0 (IOT) ±0.5% of mean value