missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 10.00, Chem-Lab

Designed for pH measurement and maintenance in moderately alkaline samples. Buffer Solution, pH 10.00, Chem-Lab is a solution of boric acid, sodium hydroxide and potassium chloride, with a pH level of 10.00 ±0.02 at 20°C. Solution is traceable to standard reference materials (SRM) from NIST.

Brand:  Chem Lab CL03.0204.1000

Product Code. 10376312

  • 33.30€ / 1 L

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

pH Levels ± 0.02:
  • pH 10.24 — 0°C
  • pH 10.10 — 10°C
  • pH 10.00 — 20°C
  • pH 9.95 — 25°C
  • pH 9.90 — 30°C
  • pH 9.82 — 40°C
  • pH 9.75 — 50°C
  • pH 9.68 — 60°C
  • pH 9.62 — 70°C
  • pH 9.55 — 80°C
  • pH 9.49 — 90°C
Specifications

Specifications

10043-35-3, 7447-40-7, 1310-73-2
100°C
1.01g/mL
10.00 ±0.02
1000 mL
Boric Acid (H3BO3), Potassium Chloride (KCl) and Sodium Hydroxide (NaOH)
Polyethylene
Soluble in water
0°C
Colorless
Laboratory use, pH measurements
Liquid
pH Buffer
Odorless
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.