missing translation for 'onlineSavingsMsg'
Learn More

pH 10.01 Calibration Buffer Sachets, Hanna Instruments™

Product Code. 11657020
Click to view available options
Quantity:
20 mL
Packaging:
25Pack
This item is not returnable. View return policy
This item is not returnable. View return policy

Premium quality pH 10.01 calibration buffer that is NIST traceable, supplied with a Certificate of Analysis, and produced in accordance with ISO 3696/BS3978 standards.

  • Premium quality pH 10.01 calibration buffer that is NIST traceable, supplied with a Certificate of Analysis, and produced in accordance with ISO 3696/BS3978 standards 
  • Uses high purity salts, deionized water, certified weight-checked balances, and Class A glassware in a temperature controlled environment monitored with certified thermometers
  • Reported values are accurate to +/- 0.01 pH @ 25°C and are traceable to NIST Standard Reference Materials (SRMs)

Specifications

Color Undesignated
pH 10.01
Physical Form Liquid
Quantity 20 mL
Formulation Aqueous Buffer Solution
Odor Odorless
Packaging 25Pack
Traceability to NIST Yes
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'