missing translation for 'onlineSavingsMsg'
Learn More

pH 4.00 Buffer Solution at 20°C, Reagecon
BIOPHARMA

Tested, certified, NIST traceable, and shelf-stable buffer solution.

Brand:  Reagecon 5040

Product Code. 11410459

  • 85.75€ / 5 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • pH Buffer Reference Standard, pH Buffer Reference Material
  • Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
  • Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications

Specifications

Accuracy ± 0.01 pH units
Liquid
HDPE Bottle
4.00 ±0.01
5 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number424L1 Certificate TypeCertificate of Analysis Date25/02/2025 Catalog Number5040
Lot Number424H1 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number5040
Lot Number424F2 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number5040
Lot Number424F1 Certificate TypeCertificate of Analysis Date25/06/2024 Catalog Number5040
Lot Number424C1 Certificate TypeCertificate of Analysis Date14/05/2024 Catalog Number5040
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
pH 4.00 Buffer Solution at 20°C, Reagecon > 5 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.