missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFI 1, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent BET bromodomain inhibitor; exhibits inhibitory activity at bromodomain-containing protein (BRD) 2 and BRD4 (IC50 values are 98 and 220 nM respectively). Induces apoptosis and G1 cell cycle arrest in BET inhibitor-sensitive cell lines (MV4;11). Also downregulates Aurora B expression in MV4;11 cells. Cell permeable.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 347.39 |
Percent Purity | >98% |
Chemical Name or Material | PFI 1 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'