missing translation for 'onlineSavingsMsg'
Learn More

PF 04217903 mesylate, Tocris Bioscience™

Product Code. 18094102
Click to view available options
10mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Highly selective MET inhibitor

Highly selective, high affinity MET inhibitor (Ki = 6-7 nM against wild type c-Met). Displays >1000-fold selectivity for c-Met over a panel of 208 kinases.

Specifications

CAS 956906-93-7
Molecular Formula C20H20N8O4S
Quantity 10 mg
Synonym pf 04217903 mesylate, pf-04217903 methanesulfonate, pf-04217903 mesylate, 2-4-3-quinolin-6-ylmethyl-1,2,3 triazolo 4,5-b pyrazin-5-yl pyrazol-1-yl ethanol; methanesulfonic acid, c19h16n8o.ch4o3s, 1h-pyrazole-1-ethanol, 4-1-6-quinolinylmethyl-1h-1,2,3-triazolo 4,5-b pyrazin-6-yl-, methanesulfonate 1:1, 2-4-1-quinolin-6-ylmethyl-1h-1,2,3 triazolo 4,5-b pyrazin-6-yl-1h-pyrazol-1-yl ethanol methanesulfonate 1:1, pf-04217903 mesylate/pf04217903 mesylate, pf hplc
InChI Key HBEMHKVWZJTVOC-UHFFFAOYSA-N
SMILES CS(=O)(=O)O.C1=CC2=C(C=CC(=C2)CN3C4=NC(=CN=C4N=N3)C5=CN(N=C5)CCO)N=C1
IUPAC Name methanesulfonic acid;2-[4-[3-(quinolin-6-ylmethyl)triazolo[4,5-b]pyrazin-5-yl]pyrazol-1-yl]ethanol
Molecular Weight (g/mol) 468.492
PubChem CID 24852079
Formula Weight 468.49
Percent Purity >98%
Chemical Name or Material PF 04217903 mesylate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'