Learn More
Isofield™ Peta 230 Non-Sterile Earloop Cleanroom Facemasks with Clip - ISO Class 4/5 Compatible
Isofield Peta is ISO Class 4 compatible. The Peta is also fitted with a convenient clip to adapt to your SOP. The individually packed Peta has excellent bacterial and particle filtration efficiency, and an excellent choice for GMP Grade C/D environments.
Brand: Isofield™ 60210
238.33 EUR valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
- Non-sterile earloop cleanroom facemask with clip
- 230 mm length with malleable nose strip
- 3-layer non-woven configuration
- Highly efficient bacterial and particle filtration
- Particle Shedding (Helmke Drum) Category I
- Individually packed to maintain low CFU count
- Pouches printed with IPA resistant ink to maintain traceability
Specifications
Cleanroom Face Mask | |
Earloops | |
Wide | |
Pleated |
White | |
Polyethylene, Polypropylene | |
Non-sterile |