missing translation for 'onlineSavingsMsg'
Learn More

Permeabilizing Solution 2 (RUO/GMP), BD Biosciences™

For the permeabilization of lymphocyte membranes prior to intracellular immunofluorescence staining with monoclonal antibodies.

Brand:  BD Biosciences 340973

Additional Details : Weight : 0.05300kg


Product Code. 15855948

  • 795.00€ / Each
Estimated Shipment: 22-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Premixed concentrate formulated specifically for use in flow cytometry.
  • For the permeabilization of lymphocyte membranes prior to intracellular immunofluorescence staining with monoclonal antibodies.
  • This formulation is designed for antigen-specific activation applications.
Specifications

Specifications

10X
Permeabilization of lymphocyte membranes prior to intracellular immunofluorescence staining with monoclonal antibodies
Flow Cytometer
500 Tests
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BD FACS Permeabilizing Solution 2

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H371-May cause damage to organs.
  • H373-May cause damage to organs.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : diethylene glycol, formaldehyde, methanol

SDS
Documents

Documents

Certificates
Promotions

Promotions