Learn More
Brady™ PermaSleeve Wiremarker Sleeves for BMP41, BMP51, and BMP53
PermaSleeve Wiremarkers Sleeves provide a wire marking solution combining speed, ease of use, and permanence to create high quality labels.
Brand:  Brady™ 143231
 View more versions of this product
Product Code. 11922762
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
PermaSleeve markers provide a wire marking solution that combines speed, ease of use, and permanence to create long-lasting, high quality identification.
  • Self-extinguishing
  • Meet the requirements of SAE AMS-DTL-23053/5 Class 1, SAE-AS-8153, an dMIL-STD-202 method 215K
  • UL224 recognized
Specifications
Specifications
Black on Yellow
Polyolefin
0.24 in.
6 mm
6 mm
2011/65/EU and 2015/863
BMP41 Label Printer, BMP51 Label Printer, BMP53 Label Printer
0.08 in.
2 mm
0.24 in.
Cartridge, Ribbon
Electrical Labeling, Wire and Cable Labeling