Learn More
Axygen™ PerkinElmer™ Janus™ & Multiprobe Robotic Tips
Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
20 μL
25 μL
50 μL
175 μL
200 μL
Sterility:
Non-sterile
Sterile
Color:
Black
Clear
Unit Size:
Pack of 4800
Pack of 960
Description
- Racked
- Black and Clear tips available
- With Liquid level sensing
- Nonsterile and Presterilized tips
- Blue, Red and Green rack color inserts
Order Info
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Specifications
Specifications
| Volume (Metric) | 200 μL |
| Filtered | Non-filtered |
| Sterility | Sterile |
| Color | Clear |
| Autoclavable | Autoclavable |
| Clean Claims | DNase, RNase, and Pyrogen free |
| Format | Rack |
| Tip Style | Automation, Round |
| Packaging | SBS Rack |
| Quantity | 50 Racks/Cs. |
| Show More |
Product Content Correction
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido