missing translation for 'onlineSavingsMsg'
Learn More

Pepsin/Hydrochloric Acid Electrode Cleaning Solution, SI Analytics™

SI Analytics™ Pepsin/Hydrochloric Acid Electrode Cleaning Solution is a corrosive solution that is suitable for cleaning electrodes.

Brand:  SI Analytics 285138295


 View more versions of this product

Product Code. 11904759

  • 34.00€ / 250mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • 250mL L5104 cleaning solution
  • Cleaning solutions for combination electrodes and reference electrodes
  • Pepsin/hydrochloric acid solution 250 ml in DURAN™ glass bottle
TRUSTED_SUSTAINABILITY
Specifications

Specifications

100°C
Cleaning Solution
1
Fully miscible in water
Colorless
Odorless
250 mL
23hPa
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Pepsin/hydrochloric acid solution

Signal Word
  • Warning

Hazard Category
  • Substances and mixtures corrosive to metals Category 1

Hazard Statement
  • H290-May be corrosive to metals.
  • EUH208-Contains . May produce an allergic reaction.

Precautionary Statement
  • P234-Keep only in original container.

Supplemental information
  • MIXTURE LIST-Contains : Pepsin A

SDS
Documents

Documents

Certificates
Promotions

Promotions