missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Pepsin Cleaning Solution
Ideal for removing protein precipitation from the diaphragm of electrodes with liquid electrolyte.
Brand: WTW™ 109648
67.58 EUR valid until 2024-12-20
Use promo code "21644" to get your promotional price.
This item is not returnable.
View return policy
Description
- Can remove protein precipitation from the diaphragm of electrodes with liquid electrolyte
- Solutions for operation, cleaning and storage of electrodes - simple refilling and safe storage warranted
- Electrolytes are used as refill solutions for liquid filled electrodes as well as a storage solution for all types of pH electrodes
Specifications
Cleaning | |
3 x 250 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- PEP/pH
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P234-Keep only in original container.
Supplemental information
- MIXTURE LIST-Contains : Pepsin A