missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 2-pyrid-3-yl-1,3-thiazole-4-carboxylate, 97%, Thermo Scientific™

Product Code. 10306544
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 906352-58-7
Molecular Formula C15H5F5N2O2S
Molecular Weight (g/mol) 372.27
MDL Number MFCD09025821
InChI Key LHRLGPDYSCUADI-UHFFFAOYSA-N
Synonym pentafluorophenyl 2-pyrid-3-yl-1,3-thiazole-4-carboxylate, pentafluorophenyl 2-pyridin-3-yl-1,3-thiazole-4-carboxylate, 2,3,4,5,6-pentafluorophenyl 2-pyridin-3-yl-1,3-thiazole-4-carboxylate, perfluorophenyl 2-pyridin-3-yl thiazole-4-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl 2-pyridin-3-yl-1,3-thiazole-4-carboxylate, 2-3-pyridinyl-4-thiazolecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 18525705
SMILES FC1=C(F)C(F)=C(OC(=O)C2=CSC(=N2)C2=CN=CC=C2)C(F)=C1F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 2-pyridin-3-yl-1,3-thiazole-4-carboxylate
Formula Weight 372.27
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 2-pyrid-3-yl-1, 3-thiazole-4-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.