Learn More
3M™ PELTOR™ WorkTunes™ Pro AM/FM Radio Headset
3M™ PELTOR™ WorkTunes™ Pro AM/FM Radio Headset helps protect you from potentially harmful workplace noise while at the same time enjoy listening to the built in AM/FM radio.
Brand:  3M™ 7100089368
 View more versions of this product
Product Code. 15740098
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Protect your hearing and listen to AM/FM radio at the same time with built-in antenna
  • Digital station search and voice guided menu system with announced frequencies
  • 3.5 mm listen only stereo input (limited to 82 dB) for connection to external devices (e.g. mobile phone, radio, two-way radio and iPod)
  • Auto power off turns headset off after 4 hours of non-use, to save energy and a low-battery warning informs you when to change batteries
  • To reduce the corrosion caused by sweat the electronics are located in the outer part of the cup
  • Store up to 5 stations on the headset
  • Available in both headband and helmet attached versions (black)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Radio Headset
HRXS221A
32 dB