Learn More
Thermo Scientific™ PEEK Fingertight Fittings
Product Code. 15319849 Shop All Thermo Scientific Products
Click to view available options
Type:
Fingertight Fitting
One-Piece PEEK Fingertight Fitting
One-Piece PEEK Long Fingertight Fitting
PEEK Wing Nut w/PEEK Ferrule
Replacement Ferrule
Short Head Port Plug, Black
Short Head Port Plug, Red
Description:
1/16 in.
1/16 in., 0.37 in. Head
1/16 in., 10-32 Coned, Delrin, Black
1/16 in., 10-32 Coned, Delrin, Red
PEEK
PEEK, 1/32 in., 0.25 in. Head
Unit Size:
Each
Pack of 10
Product Code. 15319849
Brand: Thermo Scientific™ U467R
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Ensure HPLC quality and robustness with Thermo Scientific™ PEEK Fingertight Fittings.
Ensure HPLC quality and robustness with Thermo Scientific™ PEEK Fingertight Fittings. Machined for reliability, these fittings resist cracking, breaking, thread stripping and leaking in both low- and high-pressure applications. Fingertight plumbing ensures ease of use. The fittings are biocompatible for use across a broad range of applications.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Type Short Head Port Plug, Red
Description 1/16 in., 10-32 Coned, Delrin, Red
For Use With (Equipment) 10-32 Coned Port
Material Delrin Acetal
Quantity 1 Each
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.