Learn More
Cytiva PEEK ETFE Sample Tubing Kit
Brand:  Cytiva 18111577
Product Code. 12471080
  • 207.00€ / 1 set
Estimated Shipment: 15-04-2025
to see stock.
Explore more special offers
Specifications
3/16 in.
1 mm
2 Pack
Product Title
Cytiva PEEK ETFE Sample Tubing Kit > I.D.: 1mm