Learn More
BRAND™ PCR 12-Strip Caps
Manufactured from polypropyle without lubricants or chemical additives that can contaminate reagents and samples.PCR 12-Strip Caps are autoclavable and RNase-, DNase-, and DNA-free. Caps are domed and available in a variety of colors for sample identification.
Brand:  BRAND™ 781290
 View more versions of this product
Product Code. 10080902
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
Manufactured from high-quality virgin plastics for excellent, reproducible results in most PCR applications, and are manufactured without lubricants or chemical additives that can contaminate reagents and samples. 
  • Autoclavable
  • RNase-, DNase-, and DNA-free
  • All 12-Strip caps are domed and available in a variety of colors for sample identification
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Domed
Yes
Autoclavable
PCR tubes
Polypropylene
Yes
Tube Strip Cap