missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCBD2 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97231APC
This item is not returnable.
View return policy
Description
PCBD2 Polyclonal antibody specifically detects PCBD2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PCBD2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| 4-alpha-hydroxy-tetrahydropterin dehydratase 2, DCOH2DCOHMPHS2,6-pyruvoyl-tetrahydropterin synthase/dimerization cofactor of hepatocytenuclear factor 1 alpha (TCF1) 2, DcoH-like protein DCoHm, dimerization cofactor of hepatocyte nuclear factor 1 (HNF1) from muscle, dimerization cofactor of hepatocyte nuclear factor 1 alpha (TCF1), Dimerization cofactor of hepatocyte nuclear factor 1 from muscle, EC 4.2.1.96, HNF-1-alpha dimerization cofactor, PHS 2, pterin-4 alpha-carbinolamine dehydratase 2, pterin-4 alpha-carbinolamine dehydratase/dimerization cofactor of hepatocytenuclear factor 1 alpha (TCF1) 2, pterin-4-alpha-carbinolamine dehydratase 2 | |
| Produced in rabbits immunized with E. coli-derived Human PCBD2 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 84105 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction