Learn More
Pax3/Pax7 Antibody (274212), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB2457I
Product Code. 18503788
  • 724.00€ / 0.10mL
Description
Pax3/Pax7 Monoclonal antibody specifically detects Pax3/Pax7 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pax3/Pax7
Monoclonal
Janelia Fluor 549
50mM Sodium Borate
E. coli-derived recombinant human Pax3 (isoform Pax3a), Met1-Ser215, Accession # NP_000429
0.1 mL
Apoptosis, Mesenchymal Stem Cell Markers, Stem Cell Markers, Transcription Factors and Regulators
Human, Mouse
Purified
Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
274212
Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Store at 4°C in the dark.
IgG2a