missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Patulin, 99%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Packaging:
Glass bottle
Unit Size:
10mg
25mg
5mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 108.0°C to 111.0°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (HPLC) |
| Packaging | Glass bottle |
| Quantity | 25 mg |
| Merck Index | 15, 7158 |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in most common organic solvents,soluble in ethyl and amyl acetate,insoluble in petroleum ether |
| Formula Weight | 154.12 |
| Percent Purity | 99% |
| Show More |
Safety and Handling
Product Identifier
- Patulin
- Danger
- Acute toxicity Category 3
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction